Labconco Filtermate Portable Exhausters
Use these air filtration units to pull air-borne fumes and powder through your XPert Balance Enclosure or XVS Ventilation Station. Filter removes contaminants before returning air to room or exhausting to outside. Specify efficient, high-load filters to remove general organics, particulates (HEPA filter), or both. Adjust airflow to give optimal performance. Order optional filters for formaldehyde and ammonia. HEPA filter models are available for external exhaust using a thimble connection.
FilterMate Exhausters have compact, corrosion-resistant housings of white and gray epoxy-coated steel. Rubber feet. Variable speed blower operation is quiet with a unique perforated exhaust cover. Includes IEC 3-wire electrical cord with plug.
827,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Width (in) Filter Type Power (VAC)
Availability Pricing
Labconco Filtermate Portable Exhauster, HEPA Filter, Internal Exhaust; 230V 0910062 Mfr # 3970020
COMPARE
14 HEPA 230
Call For Price
Labconco Filtermate Portable Exhauster, Carbon-Based Filter, Internal Exhaust; 230V 0910067 Mfr # 3970021
COMPARE
14 Carbon 230
₹827,000.00INR / Each
Labconco Filtermate Portable Exhauster, HEPA Filter, External Exhaust; 230V 0910072 Mfr # 3970022
COMPARE
18.625 HEPA 230
Call For Price
Labconco Filtermate Portable Exhauster, HEPA/Carbon Filters, Internal; 230V 0910077 Mfr # 3970023
COMPARE
15 HEPA/Carbon 230