Lab-Crest 110-809-0001 Insert for Coupling Component, PFA
Representative image only.
Lab-Crest 110-809-0001 Insert for Coupling Component, PFA
Lab-Crest | Mfr: 110-809-0001| Item: 6340078
Optimize performance of your Lab-Crest glassware
₹16,000.00INR / Each
₹16,000.00INR / Each
Buy It Together
Current Item
Lab-Crest 110-809-0001 Insert for Coupling Component, PFA
Lab-Crest 110-809-0001 Insert for Coupling Component, PFA
6340078
₹16,000.00INR / Each
+
Lab-Crest 110-601-0001 Female Coupling Component, Aluminum
6340064
₹21,900.00INR / Each
=
Total Price 37900.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • PFA construction—use for high-temperature applications
More About this Item
A complete coupling consists of a female coupling (63400-64 or 63400-72), male coupling (63400-66), insert (63400-68 or 63400-78), and an O-ring.
Specifications
  • MaterialPFA
  • Compatible WithLab-Crest pressure reaction vessels
  • DescriptionInsert for Coupling Component, PFA