Lab-Crest Inserts for Pressure Reaction Vessel Coupling Assembly
Optimize performance of your Lab-Crest glassware
A complete coupling consists of a female coupling (63400-64 or 63400-72), male coupling (63400-66), insert (63400-68 or 63400-78), and an O-ring.
9,500.00 - ₹16,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Material
Availability Pricing
Lab-Crest 110-759-0001 Insert for Coupling Component, PE 6340068 Mfr # 110-759-0001
COMPARE
Polyethylene (PE)
₹9,500.00INR / Each
Lab-Crest 110-809-0001 Insert for Coupling Component, PFA 6340078 Mfr # 110-809-0001
COMPARE
PFA
₹16,000.00INR / Each