Lab-Crest 110-601-0002 Female Coupling Component, SS
Representative image only.
Lab-Crest 110-601-0002 Female Coupling Component, SS
Lab-Crest | Mfr: 110-601-0002| Item: 6340072
Optimize performance of your Lab-Crest glassware
₹37,900.00INR / Each
₹37,900.00INR / Each
Buy It Together
Current Item
Lab-Crest 110-601-0002 Female Coupling Component, SS
Lab-Crest 110-601-0002 Female Coupling Component, SS
6340072
₹37,900.00INR / Each
+
Lab-Crest 110-650-0001 Male Coupling Component, SS
6340066
₹67,700.00INR / Each
+ =
Total Price 172700.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Stainless steel construction
More About this Item
A complete coupling consists of a female coupling (63400-64 or 63400-72), male coupling (63400-66), insert (63400-68 or 63400-78), and an O-ring.
Specifications
  • MaterialStainless steel
  • Compatible WithPressure reaction vessels
  • TypeFemale Coupling
  • DescriptionFemale Coupling Component, SS