Lab-Crest 110-908-0001 O-Ring for Coupling Component, PFA
Representative image only.
Lab-Crest 110-908-0001 O-Ring for Coupling Component, PFA
Lab-Crest | Mfr: 110-908-0001| Item: 6340074
Optimize performance of your Lab-Crest pressure reaction vessels
₹2,800.00INR / Each
₹2,800.00INR / Each
Buy It Together
Current Item
Lab-Crest 110-908-0001 O-Ring for Coupling Component, PFA
Lab-Crest 110-908-0001 O-Ring for Coupling Component, PFA
6340074
₹2,800.00INR / Each
+
Lab-Crest 110-601-0001 Female Coupling Component, Aluminum
6340064
₹21,900.00INR / Each
=
Total Price 24700.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • O-rings ensure a tight seal
  • PFA construction—use for high-temperature applications
More About this Item
Compatible for use with your Lab-Crest pressure reaction vessels.
A complete coupling consists of a female coupling (63400-64 or 63400-72), male coupling (63400-66), insert (63400-68 or 63400-78), and an o-ring.
Specifications
  • MaterialPFA
  • Compatible WithPressure reaction vessels
  • DescriptionO-Ring for Coupling Component, PFA