Idex Back Pressure Regulator Cartridge, 100 psi; 1/EA

Representative image only.

Idex Back Pressure Regulator Cartridge, 100 psi; 1/EA

Idex | Mfr: P-763| Item: 0201441

Select from preset pressure settings

₹27,300.00INR / Each

₹27,300.00INR / Each

Buy It Together

Current Item

Idex Back Pressure Regulator Cartridge, 100 psi; 1/EA
Idex Back Pressure Regulator Cartridge, 100 psi; 1/EA
0201441

₹27,300.00INR / Each
+ =
Total Price 81700.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Use with BPR holders
  • Red end cap, tan body
  • 125 µL swept volume
  • 1 month warranty

More About this Item

Use BPR cartridges with the holders to enhance system performance through outgassing prevention and improved pump check valve efficiency.

1 Month Warranty
Specifications
  • Connection TypeHPLC/Chromatography
  • Pressure Setting (PSI)100
  • DescriptionIdex P-763 Back Pressure Regulator Cartridge, 100 psi; 1/EA
Reviews
Q&A