Eppendorf 3124000016 Single-Channel Pipette, Gray; Fixed 10 uL
Representative image only.
Eppendorf 3124000016 Single-Channel Pipette, Gray; Fixed 10 uL
Eppendorf | Mfr: 3124000016| Item: 2450518
Provide high reproducibility with your dispensing and reduce the fatigue associated with constant repetitive thumb and arm movements.
₹206,000.00INR / Each
for an additional
₹14,500.00INR / Each
for an additional
₹25,800.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹206,000.00INR / Each
Buy It Together
Current Item
Eppendorf 3124000016 Single-Channel Pipette, Gray; Fixed 10 uL
Eppendorf 3124000016 Single-Channel Pipette, Gray; Fixed 10 uL
2450518
₹206,000.00INR / Each
+ + +
Eppendorf 022491555 50 to 1000µL epTIPS pipette tips, Reloads
1888876
₹52,900.00INR / Case of 960
+
Cole-Parmer Adjustable-Volume Pipettor, 500 to 5000 µL
2450109
₹8,400.00INR / Each
+ =
Total Price 603800.00 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Requires up to 50% less pipetting force, designed to minimize stress and fatigue
  • The ultra light pipette is designed to fit comfortably in your hand
  • Made with industry leading materials for exceptional durability
  • 3 year warranty
More About this Item
The Research Plus was optimized for form and function. The ultra lightweight pipette meets the highest needs for precision and accuracy- combined with ultimate ergonomics and increased flexibility. The spring loaded tip cone achieves optimal tighntess and minimal attachment forces-doing most of the work for you. Make your volume adjustments with just one hand- only a few turns will get you to your desired volume. The four-digit volume display is easy to view. Dissassemble your pipettor with ease- the quick connection clip lets you remove the lower part in just 2 steps. The secondary adjustment gives you greater accuracy for specific liquid types.
Safe, robust and ultralight pipettor is made from Fortron®, which is resistant to acids, alkalies, mildew, bleaches,aging, sunlight,heat, and abrasion. Fully autoclavable for simple decontamination.
3 Year Warranty
Specifications
  • SterileNo
  • Max Volume (µL)10
  • Max Volume (mL)0.01
  • Max Dispensing (mL)0.01
  • Color CodeGray
  • DescriptionSingle-Channel Pipette, Gray; Fixed 10 uL