Eppendorf epTIPS Quality Pipette Tips
Refill system and reduced ejection force requirement
  • Contamination-free transfer of trays to the working box
  • System optimized for use with multichannel pipettes
  • Color-coded trays for simple identification of tips and matching Eppendorf® pipette
  • Tips can be attached to the pipette from the refill trays
  • Refill trays are entirely autoclavable for future use; space-saving packaging
The Eppendorf epTIPS - Totally Integrated Pipetting System – satisfiy the stringent requirements of dependability, reproducibility, and the trend towards smaller sample volumes.
Optimized cone geometry allows for a perfect matching of every Eppendorf pipette tip to every Eppendorf pipette, for guaranteed pipette performance when used as a system. Additionally, the epTIPS are also universally applicable for use with pipettes from all other manufacturers.
The epTIPS system provides an alternative to throwing away an entire box when the tips run out. The Reload set enables contamination-free transfer of fresh tip trays into the reusable box used for loading your pipettor. To begin using this sytem, order a Set, which includes a reusable box and five trays of 96 tips (five trays of 24 tips for 18888-64). For one-time use tip boxes, order a Rack.
12,000.00 - ₹25,000.00INR / Case of 960
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Min Volume (µL) Package Of
Availability Pricing
Eppendorf 022491351 Refillable Box of Pipette Tips, 50 to 1000 uL, 96/bx WW-18888-42 Mfr # 0030076176
COMPARE
1000 50 Refillable Box; 96/bx
₹12,000.00INR / Case of 96
Eppendorf 022491431 2 to 200µL epTIPS pipette tips, Starter set WW-18888-56 Mfr # 0030076320
COMPARE
200 2 Refillable Box with 5 Refills; 480/pk
₹16,700.00INR / Set of 480
Eppendorf 022491504 0.1 to 10µL epTIPS pipette tips, Reloads WW-18888-66 Mfr # 022491504
COMPARE
10 0.1 10 Reloads; 960/cs
₹25,000.00INR / Case of 960
Eppendorf 022491512 0.1 to 20µL epTIPS pipette tips, Reloads WW-18888-68 Mfr # 022491512
COMPARE
20 0.1 10 Reloads; 960/cs
₹25,000.00INR / Case of 960
Eppendorf EPTIPS epTIPS Reload Trays, 0.5-20 uL; 10 trays WW-18888-70 Mfr # 022491521
COMPARE
20 0.5 10 Reloads; 960/cs
₹25,000.00INR / Case of 960
Eppendorf 022491539 2 to 200µL epTIPS pipette tips, Reloads WW-18888-72 Mfr # 022491539
COMPARE
200 2 10 Reloads; 960/cs
₹19,100.00INR / Case of 960
Eppendorf 022491547 20 to 300µL epTIPS pipette tips, Reloads WW-18888-74 Mfr # 022491547
COMPARE
300 20 10 Reloads; 960/cs
₹19,100.00INR / Case of 960
Eppendorf 022491555 50 to 1000µL epTIPS pipette tips, Reloads WW-18888-76 Mfr # 2249 1555
COMPARE
1000 50 10 Reloads; 960/cs
₹21,500.00INR / Case of 960
Eppendorf 022491563 50 to 1250µL epTIPS pipette tips, Reloads WW-18888-78 Mfr # 022491563
COMPARE
1250 50 10 Reloads; 960/cs
₹21,500.00INR / Case of 960
Eppendorf 022491571 500 to 2500µL epTIPS pipette tips, Reloads WW-18888-80 Mfr # 022491571
COMPARE
2500 500 10 Reloads; 480/cs
₹21,500.00INR / Case of 960
Eppendorf 022492012 0.1 to 20µL epTIPS pipette tips, bulk pack of 1000 WW-18889-00 Mfr # 022492012
COMPARE
20 0.1 Bulk; 1000/pk
₹20,700.00INR / Case of 1000
Eppendorf 022492047 20 to 300µL epTIPS pipette tips, bulk pack of 1000 WW-18889-06 Mfr # 022492047
COMPARE
300 20 Bulk; 1000/pk
₹16,300.00INR / Case of 1000
Eppendorf 022492055 50 to 1000µL epTIPS pipette tips,bulk pack of 1000 WW-18889-08 Mfr # 022492055
COMPARE
1000 50 Bulk; 1000/pk
₹18,100.00INR / Case of 1000
Eppendorf 022492063 50 to 1250µL epTIPS pipette tips, bulk pack of 1000 WW-18889-10 Mfr # 022492063
COMPARE
1250 50 Bulk; 1000/pk
₹18,100.00INR / Case of 1000
Eppendorf 022492071 500 to 2500µL epTIPS pipette tips, bulk pack of 500 WW-18889-12 Mfr # 022492071
COMPARE
2500 500 Bulk; 500/pk
₹20,700.00INR / Case of 500
Showing 1- 15 of 15