Eppendorf 022491547 20 to 300µL epTIPS pipette tips, Reloads
Representative image only.
Eppendorf 022491547 20 to 300µL epTIPS pipette tips, Reloads
Eppendorf | Mfr: 022491547| Item: 1888874
Refill system and reduced ejection force requirement
₹47,100.00INR / Case of 960
for an additional
₹14,500.00INR / Each
for an additional
₹25,800.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹47,100.00INR / Case of 960
Buy It Together
Current Item
Eppendorf 022491547 20 to 300µL epTIPS pipette tips, Reloads
Eppendorf 022491547 20 to 300µL epTIPS pipette tips, Reloads
1888874
₹47,100.00INR / Case of 960
+ + + =
Total Price 565600.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Contamination-free transfer of trays to the working box
  • System optimized for use with multichannel pipettes
  • Color-coded trays for simple identification of tips and matching Eppendorf® pipette
  • Tips can be attached to the pipette from the refill trays
  • Refill trays are entirely autoclavable for future use; space-saving packaging
More About this Item
The Eppendorf epTIPS - Totally Integrated Pipetting System – satisfiy the stringent requirements of dependability, reproducibility, and the trend towards smaller sample volumes.
Optimized cone geometry allows for a perfect matching of every Eppendorf pipette tip to every Eppendorf pipette, for guaranteed pipette performance when used as a system. Additionally, the epTIPS are also universally applicable for use with pipettes from all other manufacturers.
The epTIPS system provides an alternative to throwing away an entire box when the tips run out. The Reload set enables contamination-free transfer of fresh tip trays into the reusable box used for loading your pipettor. To begin using this sytem, order a Set, which includes a reusable box and five trays of 96 tips (five trays of 24 tips for 18888-64). For one-time use tip boxes, order a Rack.
Specifications
  • SterileNo
  • Max Volume (µL)300
  • Min Volume (µL)20
  • Min Volume (mL)0.02
  • Max Volume (mL)0.3
  • Tip Volume (mL)0.3
  • Min Dispensing (mL)0.02
  • Max Dispensing (mL)0.3
  • Overall Length (InchShort)2.125
  • Color CodeOrange
  • Package Of10 Reloads; 960/cs
  • Compatible withUniversal
  • DescriptionQuality