Eppendorf Research plus Fixed Volume Pipettors
Provide high reproducibility with your dispensing and reduce the fatigue associated with constant repetitive thumb and arm movements.
  • Requires up to 50% less pipetting force, designed to minimize stress and fatigue
  • The ultra light pipette is designed to fit comfortably in your hand
  • Made with industry leading materials for exceptional durability
The Research Plus was optimized for form and function. The ultra lightweight pipette meets the highest needs for precision and accuracy- combined with ultimate ergonomics and increased flexibility. The spring loaded tip cone achieves optimal tighntess and minimal attachment forces-doing most of the work for you. Make your volume adjustments with just one hand- only a few turns will get you to your desired volume. The four-digit volume display is easy to view. Dissassemble your pipettor with ease- the quick connection clip lets you remove the lower part in just 2 steps. The secondary adjustment gives you greater accuracy for specific liquid types.
Safe, robust and ultralight pipettor is made from Fortron®, which is resistant to acids, alkalies, mildew, bleaches,aging, sunlight,heat, and abrasion. Fully autoclavable for simple decontamination.
76,300.00INR / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Code Max Volume (mL) Max Volume (µL)
Availability Pricing
Eppendorf 3124000016 Single-Channel Pipette, Gray; Fixed 10 uL WW-24505-18 Mfr # 3124000016
COMPARE
Gray 0.01 10
₹76,300.00INR / Each
Eppendorf 3124000032 Single-Channel Pipette, Light-Gray; Fixed 20 uL WW-24505-19 Mfr # 3124000032
COMPARE
Light-gray 0.02 20
₹76,300.00INR / Each
Eppendorf 3121000024 Single-Channel Pipette, Yellow; Fixed 10 uL WW-24505-21 Mfr # 3124000024
COMPARE
Yellow 0.01 10
₹76,300.00INR / Each
Eppendorf 3124000040 Single-Channel Pipette, Yellow; Fixed 20 uL WW-24505-22 Mfr # 3124000040
COMPARE
Yellow 0.02 20
₹76,300.00INR / Each
Eppendorf 3124000059 Single-Channel Pipette, Yellow; Fixed 25 uL WW-24505-23 Mfr # 3124000059
COMPARE
Yellow 0.025 25
₹76,300.00INR / Each
Eppendorf 3124000067 Single-Channel Pipette, Yellow; Fixed 50 uL WW-24505-24 Mfr # 3124000067
COMPARE
Yellow 0.05 50
₹76,300.00INR / Each
Eppendorf 3124000075 Single-Channel Pipette, Yellow; Fixed 100 uL WW-24505-25 Mfr # 3124000075
COMPARE
Yellow 0.1 100
₹76,300.00INR / Each
Eppendorf 3124000083 Single-Channel Pipette, Yellow; Fixed 200 uL WW-24505-26 Mfr # 3124000083
COMPARE
Yellow 0.2 200
₹76,300.00INR / Each
Eppendorf 3124000091 Single-Channel Pipette, Blue; Fixed 200 uL WW-24505-27 Mfr # 3124000091
COMPARE
Blue 0.2 200
₹76,300.00INR / Each
Eppendorf 3124000105 Single-Channel Pipette, Blue; Fixed 250 uL WW-24505-28 Mfr # 3124000105
COMPARE
Blue 0.25 250
₹76,300.00INR / Each
Eppendorf 3124000113 Single-Channel Pipette, Blue; Fixed 500 uL WW-24505-29 Mfr # 3124000113
COMPARE
Blue 0.5 500
₹76,300.00INR / Each
Eppendorf 3124000121 Single-Channel Pipette, Blue; Fixed 1000 uL WW-24505-31 Mfr # 3124000121
COMPARE
Blue 1 1000
₹76,300.00INR / Each
Showing 1- 12 of 12