Environmental Express Transfer Rack Set, 18 x 50 mL; 1 Rack/Each
Representative image only.
Environmental Express Transfer Rack Set, 18 x 50 mL; 1 Rack/Each
Environmental Express | Item: 5320111
Optimize performance of your Environmental Express HotBlock digestion tubes
₹14,700.00INR / Each
₹14,700.00INR / Each
Buy It Together
Current Item
Environmental Express Transfer Rack Set, 18 x 50 mL; 1 Rack/Each
Environmental Express Transfer Rack Set, 18 x 50 mL; 1 Rack/Each
5320111
₹14,700.00INR / Each
+
Environmental Express Polypropylene Storage Rack
5310004
₹12,700.00INR / Each
+ + + =
Total Price 2004300.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Watch glasses to cover your tubes during digestion
  • Transfer racks to avoid spillage
  • Caps to close tubes
  • 1 year warranty
  • Model number SC200
More About this Item
Choose from a wide range of accessories and replacement parts for your digestion tubes.
Transfer racks are constructed of clear polycarbonate and can be used for cup transfer/storage, and are designed to set directly into the compatible HotBlock and HotBlock Pro system. Handles are molded into the sides for easy placement of all digestion cups into, and removal from, the HotBlock simultaneously.
1 Year Warranty
Specifications
  • MaterialPolycarbonate (PC)
  • Compatible With50 mL digestion tubes
  • DescriptionTransfer Rack Set, 18 x 50 mL; 1 Rack/Each