Environmental Express SC296 Transfer Rack Set, 32 x 15 mL; 3 Racks/Pk
Representative image only.
Environmental Express SC296 Transfer Rack Set, 32 x 15 mL; 3 Racks/Pk
Environmental Express | Item: 5320549
Optimize performance of your Environmental Express HotBlock digestion tubes
Call For Price
Call For Price
Buy It Together
Current Item
Environmental Express SC296 Transfer Rack Set, 32 x 15 mL; 3 Racks/Pk
Environmental Express SC296 Transfer Rack Set, 32 x 15 mL; 3 Racks/Pk
5320549
Call For Price
+
Environmental Express Polypropylene Storage Rack
5310004
₹12,700.00INR / Each
+ + + =
Total Price 1989600.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Watch glasses to cover your tubes during digestion
  • Transfer racks to avoid spillage
  • Caps to close tubes
  • Model number SC296
More About this Item
Choose from a wide range of accessories and replacement parts for your digestion tubes.
Transfer racks are constructed of clear polycarbonate and can be used for cup transfer/storage, and are designed to set directly into the compatible HotBlock, HotBlock TKN, and HotBlock Pro system. Handles are molded into the sides for easy placement of all digestion cups into, and removal from, the HotBlock simultaneously.
Specifications
  • MaterialPolycarbonate (PC)
  • Compatible With15 mL digestion tubes
  • DescriptionTransfer Rack Set, 32 x 15 mL; 3 Racks/Pk