Environmental Express SC225 Transfer Rack Set, 35 x 100 mL; 3 Racks/Pk
Representative image only.
Environmental Express SC225 Transfer Rack Set, 35 x 100 mL; 3 Racks/Pk
Environmental Express | Item: 5320548
Optimize performance of your Environmental Express HotBlock digestion tubes
Call For Price
Call For Price
Buy It Together
Current Item
Environmental Express SC225 Transfer Rack Set, 35 x 100 mL; 3 Racks/Pk
Environmental Express SC225 Transfer Rack Set, 35 x 100 mL; 3 Racks/Pk
5320548
Call For Price
+
Environmental Express Polypropylene Storage Rack
5310004
₹12,700.00INR / Each
+ + + =
Total Price 2097600.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Watch glasses to cover your tubes during digestion
  • Transfer racks to avoid spillage
  • Caps to close tubes
  • Model number SC235
More About this Item
Choose from a wide range of accessories and replacement parts for your digestion tubes.
Transfer racks are constructed of clear polycarbonate and can be used for cup transfer/storage, and are designed to set directly into the compatible HotBlock and HotBlock Pro system. Handles are molded into the sides for easy placement of all digestion cups into, and removal from, the HotBlock simultaneously.
Specifications
  • MaterialPolycarbonate (PC)
  • Compatible With100 mL digestion tubes
  • DescriptionTransfer Rack Set, 35 x 100 mL; 3 Racks/Pk