Environmental Express Disposable Watch Glass, 100 mL; 500/Pack
Representative image only.
Environmental Express Disposable Watch Glass, 100 mL; 500/Pack
Environmental Express | Item: 5320108
Optimize performance of your Environmental Express HotBlock digestion tubes
₹29,600.00INR / Pkg of 500
₹29,600.00INR / Pkg of 500
Buy It Together
Current Item
Environmental Express Disposable Watch Glass, 100 mL; 500/Pack
Environmental Express Disposable Watch Glass, 100 mL; 500/Pack
5320108
₹29,600.00INR / Pkg of 500
+
Environmental Express Polypropylene Storage Rack
5310004
₹12,700.00INR / Each
+ + + =
Total Price 2019200.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Watch glasses to cover your tubes during digestion
  • Transfer racks to avoid spillage
  • Caps to close tubes
  • 1 year warranty
  • Model number SC610
More About this Item
Choose from a wide range of accessories and replacement parts for your digestion tubes.
Watch glasses are made from metals-free polypropylene and are ideal for digestions where reagents need to be added throughout the heating process. Watch glasses have a center stem for easy handling and to decrease the chances of contamination.
1 Year Warranty
Specifications
  • MaterialPolypropylene (PP)
  • Compatible With100 mL digestion tubes
  • DescriptionDisposable Watch Glass, 100 mL; 500/Pack