Environmental Express Disposable Reflux Caps, 50 mL; 1000/Pk

Representative image only.

Environmental Express Disposable Reflux Caps, 50 mL; 1000/Pk

Environmental Express | Item: 5320106

Optimize performance of your Environmental Express HotBlock digestion tubes

₹38,600.00INR / Pkg of 1000

₹38,600.00INR / Pkg of 1000

Buy It Together

Current Item

Environmental Express Disposable Reflux Caps, 50 mL; 1000/Pk
Environmental Express Disposable Reflux Caps, 50 mL; 1000/Pk
5320106

₹38,600.00INR / Pkg of 1000
+
Environmental Express Polypropylene Storage Rack
5310004

₹12,700.00INR / Each
+ + + =
Total Price 2028200.00 ADD 5 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Watch glasses to cover your tubes during digestion
  • Transfer racks to avoid spillage
  • Caps to close tubes
  • 1 year warranty
  • Model number SC506

More About this Item

Choose from a wide range of accessories and replacement parts for your digestion tubes.

Reflux caps are made from metals-free polypropylene and are ideal for digestions where reagents need to be added throughout the heating process. The reflux caps fit onto the 50-mL digestion cups and have a 1/2" (12.7 mm) hole in the center for easy reagent addition.

1 Year Warranty
Specifications
  • MaterialPolypropylene (PP)
  • Compatible With50 mL digestion tubes
  • DescriptionDisposable Reflux Caps, 50 mL; 1000/Pk
  • Warranty1 year
Reviews
Q&A