DWK Life Sciences (Wheaton) Staining Dish System with SS Rack and Cover; 50-Slide Capacity
Representative image only.
DWK Life Sciences (Wheaton) Staining Dish System with SS Rack and Cover; 50-Slide Capacity
DWK Life Sciences (Wheaton) | Mfr: 900400| Item: 4858510
Designed for secure staining of microscope slides
₹54,600.00INR / Each
₹54,600.00INR / Each
Buy It Together
Current Item
DWK Life Sciences (Wheaton) Staining Dish System with SS Rack and Cover; 50-Slide Capacity
DWK Life Sciences (Wheaton) Staining Dish System with SS Rack and Cover; 50-Slide Capacity
4858510
₹54,600.00INR / Each
+ + =
Total Price 97800.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Rack handle allows for convenient moving around the lab
  • Complete with dish, cover and rack
More About this Item
Choose the staining dish and rack system that meets your application requirements. Accommodate slides measuring 75 x 25 mm (3" x 1"), 75 x 38 mm (3" x 1 1⁄2"), and 75 x 51 mm (3" x 2").
This slotted rack holds 50 microscope slides, sizes 3" x 1" (75 mm x 25 mm), 3" x 1-1/2" (75 mm x 38 mm), and 3" x 2" (75 mm x 51 mm). The rack is made of non-tarnishable stainless steel that is resistant to staining solutions. The handle is permanently attached, but hinged to permit closure of the dish and easy insertion and removal of the microscope slides. The dish is manufactured from soda-lime glass. Approximate inside dimensions: 185 mm L x 88 mm W x 68 mm D.
Specifications
  • Capacity (Number of slides)50
  • DescriptionStaining Dish System with SS Rack and Cover; 50-Slide Capacity