DWK Life Sciences (Wheaton) 900170 Glass Staining Dish and Cover, without Rack; 10-Slide Capacity
Representative image only.
DWK Life Sciences (Wheaton) 900170 Glass Staining Dish and Cover, without Rack; 10-Slide Capacity
DWK Life Sciences (Wheaton) | Mfr: 900170| Item: 4858560
Designed for secure staining of microscope slides
₹9,200.00INR / Each
₹9,200.00INR / Each
Buy It Together
Current Item
DWK Life Sciences (Wheaton) 900170 Glass Staining Dish and Cover, without Rack; 10-Slide Capacity
DWK Life Sciences (Wheaton) 900170 Glass Staining Dish and Cover, without Rack; 10-Slide Capacity
4858560
₹9,200.00INR / Each
+ + =
Total Price 52400.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Rack handle allows for convenient moving around the lab
  • Holds 10 single slide or 20 slides back-to-back
  • Staining dish resists stains from Eosin and Hemotoxylin
More About this Item
Choose the staining dish and rack system that meets your application requirements. Accommodate slides measuring 75 x 25 mm (3" x 1"), 75 x 38 mm (3" x 1 1⁄2"), and 75 x 51 mm (3" x 2").
Accommodates 3 x 1" microscope slides (75 x 25mm) which slide into grooves in the glass. Made with soda lime glass which resists stains. Holds 10 single slides, or 19 slides arranged alternately straight across and diagonally, or 20 slides back to back. Approximate inside dimensions: 75 mm L x 55 mm W x 35 mm D.
Specifications
  • Capacity (Number of slides)10 to 20
  • External Width (cm)5.5
  • External Height (cm)7.5
  • External Depth (cm)3.5
  • DescriptionGlass Staining Dish and Cover, without Rack; 10-Slide Capacity