Diba Omnifit® T-Series Solvent Bottle Cap, GL38/38-430 (glass), 2 Luer Ports without Valve
Representative image only.
Diba Omnifit® T-Series Solvent Bottle Cap, GL38/38-430 (glass), 2 Luer Ports without Valve
Diba | Mfr: 00938T-2| Item: 2194290
Safe and dependable solvent delivery system without any fittings
₹5,500.00INR / Each
₹5,500.00INR / Each
Buy It Together
Current Item
Customers Also Bought
Product Description
Key Features
  • Accepts standard male Luer fittings to connect various tubing sizes
  • Anti-twist design
More About this Item
The T-series solvent bottle caps help contain vapors and protect lab staff from exposure to toxic fumes. Push through 1/8" OD semi-rigid tubing for a liquid- and vapor-tight seal or use standard male Luer fittings to connect other tubing sizes. Luer port lets you also connect a syringe for liquid removal. Anti-twist cap design allows the PTFE body to spin freely within the outer cap. No more twisted tubes and no need to disconnect fluid lines before unscrewing the cap. Caps are rated for use from -10 to 20 psi when using 1/8" OD through tubes; -10 to 0 psi when using male Luer connectors. All caps are autoclavable.
Use activated carbon vapor trap with Luer connection to prevent any dangerous solvent from escaping into the laboratory. Connect 1/16" or 1/8" ID flexible tubing or 1/16” OD semi-rigid tubing to caps with Luer adapters. Plug any unused ports with a Luer plug to ensure a closed system. Bottle thread adapter converts bottles with S40 threads to accept any GL45 thread caps.
Specifications
  • Cap MaterialPTFE inner body/PP outer cap
  • Cap / Stopper Size38-430 (GL38)
  • Cap ColorBlue
  • AutoclavableYes
  • Special Features2 x Luer
  • DescriptionDiba Omnifit® T-Series Solvent Waste Cap, GL38/38-430 (glass), 2 Luer ports without valve; 1/ea