Idex A-620 Bottle Cap Assembly, 1/8"; 1/EA
Representative image only.
Idex A-620 Bottle Cap Assembly, 1/8"; 1/EA
Idex | Mfr: A-620| Item: 0203328
Easy to use with no threaded ports or fittings required
₹9,200.00INR / Each
₹9,200.00INR / Each
Buy It Together
Current Item
Idex A-620 Bottle Cap Assembly, 1/8"; 1/EA
Idex A-620 Bottle Cap Assembly, 1/8"; 1/EA
0203328
₹9,200.00INR / Each
+ =
Total Price 14600.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • For use with tubing that has an outer diameter of 1/8"
  • Requires no threaded ports or fittings
  • 1 year warranty
More About this Item
This Bottle Cap Assembly is designed for a one-liter bottle. It has excellent resistance to solvents due to being made of ethylene tetrafluoroethylene and polypropylene.
1 Year Warranty
Specifications
  • Capacity (L)1
  • Cap Material Genuine Tefzel™ ETFE Resin, Polypropylene
  • Cap ColorRed
  • Outside Diameter (OD) (in)1/8
  • DescriptionBottle Cap Assembly, 1/8"; 1/EA