Diba Omnifit® Activated Carbon Vapor Trap for Solvent Waste Caps with Luer Ports; 1/ea
Representative image only.
Diba Omnifit® Activated Carbon Vapor Trap for Solvent Waste Caps with Luer Ports; 1/ea
Diba | Mfr: 009CF-T| Item: 2194329
Prevent chemical vapors from escaping from solvent containers
Product Description
Key Features
  • Compatible with Diba T-series solvent caps
  • Simply push-fit into a spare Luer port
More About this Item
Use traps with Luer connection with T-series solvent waste caps without valve and filter. Traps with 1/4"-28 UNF(M) connection works with caps with vent. Traps with UNF connection feature an indicator strip that changes color as it gets saturated.
Use activated carbon trap with T-series solvent caps without valve and filter. Trap contains approximately 25 mL charcoal. The frequency of exchange depends on usuage and type of solvent used.
Specifications
  • Compatible WithT-series solvent caps without valve and filter; male luer slip connection