Cole-Parmer VapLock™ Solvent Waste Manifold, PP, 83B, 1x1/2" NPT, 3x1/4" NPT; 1/EA
Representative image only.
Cole-Parmer VapLock™ Solvent Waste Manifold, PP, 83B, 1x1/2" NPT, 3x1/4" NPT; 1/EA
Cole-Parmer | Item: 1201943
Connect solvent waste manifold caps to waste containers for a secure closed system
₹31,300.00INR / Each
₹31,300.00INR / Each
Buy It Together
Current Item
Cole-Parmer VapLock™ Solvent Waste Manifold, PP, 83B, 1x1/2" NPT, 3x1/4" NPT; 1/EA
Cole-Parmer VapLock™ Solvent Waste Manifold, PP, 83B, 1x1/2" NPT, 3x1/4" NPT; 1/EA
1201943
₹31,300.00INR / Each
+ + + + + =
Total Price 124200.00 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose from polypropylene or PTFE construction
  • 83B connection
  • Polypropylene construction
  • 4 ports
More About this Item
Select from various manifold cap sizes to fit your laboratory and industrial solvent waste containers. Bottle and carboy caps are available with polypropylene (PP) or PTFE manifolds and 4 or 10 variously sized ports. Ports accept male thread adapters to connect flexible and rigid tubing, or filter 12019-79 or filter/indicator 12019-80.
Specifications
  • MaterialPolypropylene(PP)
  • Cap / Stopper Size83B
  • Cap ColorWhite
  • Special Features1 x 1/2" NPT(F), 3 x 1/4" NPT(F)
  • DescriptionSolvent Waste Manifold, PP, 83B, 1x1/2" NPT, 3x1/4" NPT; 1/EA