CPC (Colder) Twin Tube™ PTC Quick-Disconnect Fittings, Hose Barb Insert, Acetal, Non-Valved, 1/16" ID; 1/Each
Representative image only.
CPC (Colder) Twin Tube™ PTC Quick-Disconnect Fittings, Hose Barb Insert, Acetal, Non-Valved, 1/16" ID; 1/Each
CPC (Colder) | Mfr: PTC22010| Item: 5013673
Provides quick disconnect for two separate fluid lines
₹2,600.00INR / Each
₹2,600.00INR / Each
Buy It Together
Current Item
CPC (Colder) Twin Tube™ PTC Quick-Disconnect Fittings, Hose Barb Insert, Acetal, Non-Valved, 1/16" ID; 1/Each
CPC (Colder) Twin Tube™ PTC Quick-Disconnect Fittings, Hose Barb Insert, Acetal, Non-Valved, 1/16" ID; 1/Each
5013673
₹2,600.00INR / Each
+ + =
Total Price 20700.00 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
  • Two individual flow paths in one unit
  • Free coupling rotation eliminates kinked tubing
  • Ideal for tubing sizes from 1/16” to 1/8” ID
  • Flow size 3/32” (2.4 mm)
  • 1/16" hose barb
More About this Item
The CPC (Colder) Twin Tube™ PTC Series quick-disconnect hose barb body and insert fittings provide a convenient solution for managing two separate fluid lines through a single coupling. These non-valved fittings maintain individual flow paths, ensuring efficient and streamlined operation.
Featuring an ergonomic thumb latch, the fittings allow for effortless, one-handed disconnection of both lines with a single button press, even when gloves are worn.
Designed to withstand pressures from vacuum to 120 psi (8.3 bar), these fittings operate effectively within a temperature range of -40°F to 180°F (-40°C to 82°C).
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialAcetal
  • O-Ring MaterialBuna-N
  • Connection TypeQuick-Disconnect Hose Barb Insert, Non-Valved
  • Barb Size / Tubing Id (Port 1) (in)1/16
  • Flow PatternStraight
  • ColorWhite
  • Min Temperature (° F)-40
  • Max Temperature (° F)180
  • Min Temperature (° C)-40
  • Max Temperature (° C)82
  • Max Operating Pressure (PSI)120
  • DescriptionQuick-Disconnect Fittings, Hose Barb Insert, Acetal, Non-Valved, 1/16" ID