CPC® (Colder) Twin Tube™ PTC Quick-Disconnect Fittings, Hose Barb Body & Insert, Non-Valved
Provides quick disconnect for two separate fluid lines
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
  • Two individual flow paths in one unit
  • Free coupling rotation eliminates kinked tubing
  • Ideal for tubing sizes from 1/16” to 1/8” ID
  • Flow size 3/32” (2.4 mm)
The CPC (Colder) Twin Tube™ PTC Series quick-disconnect hose barb body and insert fittings provide a convenient solution for managing two separate fluid lines through a single coupling. These non-valved fittings maintain individual flow paths, ensuring efficient and streamlined operation.
Featuring an ergonomic thumb latch, the fittings allow for effortless, one-handed disconnection of both lines with a single button press, even when gloves are worn.
Designed to withstand pressures from vacuum to 120 psi (8.3 bar), these fittings operate effectively within a temperature range of -40°F to 180°F (-40°C to 82°C).
1,900.00 - ₹3,200.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Connection Type
Availability Pricing
CPC (Colder) Twin Tube™ PTC Quick-Disconnect Fittings, Hose Barb Body, Panel Mount, Acetal, Non-Valved, 1/8" ID; 1/Each 5013672 Mfr # PTC16020
COMPARE
1/8 Acetal Quick-Disconnect Hose Barb Body, Panel Mount, Non-Valved
₹3,200.00INR / Each
CPC (Colder) Twin Tube™ PTC Quick-Disconnect Fittings, Hose Barb Insert, Acetal, Non-Valved, 1/16" ID; 1/Each 5013673 Mfr # PTC22010
COMPARE
1/16 Acetal Quick-Disconnect Hose Barb Insert, Non-Valved
₹2,600.00INR / Each
CPC (Colder) Twin Tube™ PTC Quick-Disconnect Fittings, Hose Barb Insert, Acetal, Non-Valved, 1/8" ID; 1/Each 5013674 Mfr # PTC22020
COMPARE
1/8 Acetal Quick-Disconnect Hose Barb Insert, Non-Valved
₹2,000.00INR / Each
CPC (Colder) Twin Tube™ PTC Quick-Disconnect Fittings, Hose Barb Insert, ABS, Non-Valved, 1/8" ID; 1/Each 5013675 Mfr # PTC2202096
COMPARE
1/8 ABS Quick-Disconnect Hose Barb Insert, Non-Valved
₹1,900.00INR / Each