CPC® (Colder) DrumQuik® PRO Quick-Disconnect Dispensing System
Offers safe, easy, and fast container change-out of chemicals
  • Automatic shutoff valve stops flow when disconnected
  • Disposable insert reduces chemical exposure and messy handling
  • Ported vent system provides vapor management
The DrumQuik dispensing system enables safe, efficient chemical extraction from drums, jerry cans, and IBCs. Its closed-system design and durable construction meet strict chemical handling safety standards and withstand harsh environments. The system reduces chemical exposure, minimizes spills, and ensures controlled delivery from bulk containers. A ported vent allows fume control, blanket gas connection, or pressure dispensing.
The reusable coupler and cost-effective, disposable dip-tube assembly support single-use applications and can be recycled with the container. The system includes a coupler, drum insert, and dip tube (each sold separately).
920.00 - ₹13,300.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Flow Pattern Description
Availability Pricing
CPC (Colder) DrumQuik® PRO Hose Barb Fitting, Polypropylene, 1/2" NPT(F) X 1/2" ID; 1/Each 5013508 Mfr # 2403000
COMPARE
Polypropylene Straight Hose Barb Fitting, 1/2" NPT(F) X 1/2" ID
₹1,600.00INR / Each
CPC (Colder) DrumQuik® PRO Elbow Coupler, Polypropylene, with EPDM O-Rings, 316 Stainless Steel Spring, 1/2" NPT(M); 1/Each 5013515 Mfr # DQPRO120208
COMPARE
Polypropylene Elbow Elbow Coupler, Polypropylene, with EPDM O-Rings 316 SS Spring, 1/2" NPT(M)
₹13,300.00INR / Each
CPC (Colder) DrumQuik® PRO Drum Insert with 2" American Buttress Thread, HDPE and EPDM O-Rings; 1/Each 5013516 Mfr # DQPRO202BUT
COMPARE
HDPE Straight Drum Insert, High Density Polyethylene, with 2" American Buttress Thread and EPDM O-Rings
₹2,100.00INR / Each
CPC (Colder) DrumQuik® PRO Dip Tube for 55 Gal (200 L) Drums, HDPE, 35.5" (90.5 cm) Length; 1/Each 5013517 Mfr # DQPRODT0985
COMPARE
HDPE Straight Dip Tube for 55 Gal (200 L) Drums, High Density Polyethylene, 35.5" (90.5 cm) Length
₹920.00INR / Each
CPC (Colder) DrumQuik® PRO Dip Tube for 275 Gal (1040 L) Intermediate Bulk Containers, HDPE, 39.0" (99 cm) Length; 1/Each 5013518 Mfr # DQPRODT1070
COMPARE
HDPE Straight Dip Tube for 275 Gal (1040 L) Intermediate Bulk Containers, High Density Polyethylene, 39.0" (99 cm) Length
₹1,200.00INR / Each