CPC (Colder) DrumQuik® PRO Drum Insert with 2" American Buttress Thread, HDPE and EPDM O-Rings; 1/Each
Representative image only.
CPC (Colder) DrumQuik® PRO Drum Insert with 2" American Buttress Thread, HDPE and EPDM O-Rings; 1/Each
CPC (Colder) | Mfr: DQPRO202BUT| Item: 5013516
Offers safe, easy, and fast container change-out of chemicals
₹2,100.00INR / Each
₹2,100.00INR / Each
Buy It Together
Current Item
CPC (Colder) DrumQuik® PRO Drum Insert with 2" American Buttress Thread, HDPE and EPDM O-Rings; 1/Each
CPC (Colder) DrumQuik® PRO Drum Insert with 2" American Buttress Thread, HDPE and EPDM O-Rings; 1/Each
5013516
₹2,100.00INR / Each
+ + =
Total Price 8800.00 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Automatic shutoff valve stops flow when disconnected
  • Disposable insert reduces chemical exposure and messy handling
  • Ported vent system provides vapor management
More About this Item
The DrumQuik dispensing system enables safe, efficient chemical extraction from drums, jerry cans, and IBCs. Its closed-system design and durable construction meet strict chemical handling safety standards and withstand harsh environments. The system reduces chemical exposure, minimizes spills, and ensures controlled delivery from bulk containers. A ported vent allows fume control, blanket gas connection, or pressure dispensing.
The reusable coupler and cost-effective, disposable dip-tube assembly support single-use applications and can be recycled with the container. The system includes a coupler, drum insert, and dip tube (each sold separately).
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialHDPE
  • Connection TypeAmerican ButtressThreaded
  • Flow PatternStraight
  • ColorWhite
  • Min Temperature (° F)-20
  • Max Temperature (° F)120
  • Min Temperature (° C)-29
  • Max Temperature (° C)49
  • DescriptionDrum Insert, High Density Polyethylene, with 2" American Buttress Thread and EPDM O-Rings