CPC® (Colder) NS6 Quick-Disconnect Fittings, Threaded Body & Insert, Valved, Polypropylene
Designed for a pressure-activated release without leaks
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
  • Chemically resistant
  • Ideal for tubing sizes from 3/8” to 1/2” ID
The CPC (Colder) NS6 Series quick-disconnect threaded insert fittings are designed with non-spill valves to prevent spills, reduce downtime, and improve operator safety. These lightweight, compact fittings offer excellent chemical resistance and user-friendly operation.
An ergonomic thumb latch enables effortless one-handed connection and disconnection, even when wearing gloves. The fitting operates efficiently in temperatures ranging from 32 to 160 °F (0 to 71 °C) and withstands pressures from vacuum to 120 psi (8.3 bar).
3,200.00 - ₹4,800.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Flow Pattern Connection Type Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
CPC (Colder) NS6 Quick-Disconnect Fittings, Threaded Body, Polypropylene, Valved, 1/2" NPT(M); 1/Each 5013594 Mfr # NS6D10008
COMPARE
Straight Quick-Disconnect Threaded Body, Valve 1/2
₹4,800.00INR / Each
CPC (Colder) NS6 Quick-Disconnect Fittings, Threaded Insert, Polypropylene, Valved, 1/2" NPT(M); 1/Each 5013597 Mfr # NS6D24008
COMPARE
Straight Quick-Disconnect Threaded Insert, Valve 1/2
₹3,200.00INR / Each