CPC (Colder) DrumQuik® PRO Hose Barb Fitting, Polypropylene, 1/2" NPT(F) X 1/2" ID; 1/Each
Representative image only.
CPC (Colder) DrumQuik® PRO Hose Barb Fitting, Polypropylene, 1/2" NPT(F) X 1/2" ID; 1/Each
CPC (Colder) | Mfr: 2403000| Item: 5013508
Offers safe, easy, and fast container change-out of chemicals
₹1,600.00INR / Each
₹1,600.00INR / Each
Buy It Together
Current Item
CPC (Colder) DrumQuik® PRO Hose Barb Fitting, Polypropylene, 1/2" NPT(F) X 1/2" ID; 1/Each
CPC (Colder) DrumQuik® PRO Hose Barb Fitting, Polypropylene, 1/2" NPT(F) X 1/2" ID; 1/Each
5013508
₹1,600.00INR / Each
+ + =
Total Price 9600.00 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Automatic shutoff valve stops flow when disconnected
  • Disposable insert reduces chemical exposure and messy handling
  • Ported vent system provides vapor management
More About this Item
The DrumQuik dispensing system enables safe, efficient chemical extraction from drums, jerry cans, and IBCs. Its closed-system design and durable construction meet strict chemical handling safety standards and withstand harsh environments. The system reduces chemical exposure, minimizes spills, and ensures controlled delivery from bulk containers. A ported vent allows fume control, blanket gas connection, or pressure dispensing.
The reusable coupler and cost-effective, disposable dip-tube assembly support single-use applications and can be recycled with the container. The system includes a coupler, drum insert, and dip tube (each sold separately).
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPolypropylene
  • Connection TypeThreaded to Hose Barb
  • Thread Size NPT(F) (Port 1) (InchShort)1/2
  • Barb Size / Tubing Id (Port 2) (in)1/2
  • Flow PatternStraight
  • ColorBlack
  • DescriptionHose Barb Fitting, 1/2" NPT(F) X 1/2" ID