Cole-Parmer Variable Area Flowmeter, Acrylic, Air, 50 LPM, 1/8" Stainless, No Valve
Representative image only.
Cole-Parmer Variable Area Flowmeter, Acrylic, Air, 50 LPM, 1/8" Stainless, No Valve
Cole-Parmer | Item: 1530850
Our variable area acrylic flowmeters offer precision and durability for a wide range of applications
₹13,900.00INR / Each
for an additional
₹49,800.00INR / Each
for an additional
₹63,000.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹13,900.00INR / Each
Buy It Together
Current Item
Cole-Parmer Variable Area Flowmeter, Acrylic, Air, 50 LPM, 1/8" Stainless, No Valve
Cole-Parmer Variable Area Flowmeter, Acrylic, Air, 50 LPM, 1/8" Stainless, No Valve
1530850
₹13,900.00INR / Each
+
Alicat Mass Flowmeter with Display, 0 - 1 SLPM
1530881
₹169,000.00INR / Each
+
InnoCal NIST-Traceable Calibration; Air/Gas Flowmeter
1708000
₹49,800.00INR / Each
+ =
Total Price 263600.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Crystal-clear acrylic material with a stable float ensures easy viewing and long-lasting performance
  • Superior, corrosion-resistant stainless-steel fittings ensure long-term reliability in harsh environments
  • Easily convertible from panel to partial or full in-line mounting configurations to suit your needs
  • Easy-to-read English or metric scales for clear and precise measurement
  • Panel mounting with hex nuts makes installation simple and secure
  • Easy disassembly and reassembly for quick cleaning and maintenance
  • 1 year warranty
More About this Item
Whether you need an accurate air flow measurement solution for industrial processes, lab setups, or HVAC systems, our acrylic flowmeters provide the perfect combination of precision, durability, and convenience. With a variety of customizable options, they are designed to fit seamlessly into your workflow. Flowmeters without valves are best suited for applications where the flow rate remains constant or is controlled by another device.
1 Year Warranty
Specifications
  • Accuracy±5% of full scale reading
  • Max Pressure (PSI)100
  • Max Flow Rate (LPM)50
  • Max Flow Rate (SCFH)100
  • Material O-RingViton®
  • Material FloatStainless
  • Material HousingAcrylic
  • Max Operating Temperature (° F)150
  • Max Operating Temperature (° C)65
  • Process Connection (NPT(F))1/8
  • Media TypeAir
  • DescriptionVariable Area Flowmeter, Acrylic, Air, 50 LPM, 1/8" Stainless, No Valve