Cole-Parmer VapLock™ Fitting, Yellow PP w/Yellow ETFE, Straight, Compression to Threaded Adapter, 1/8" OD tubing x 1/4-28 UNF(M); 1/Ea
Representative image only.
Cole-Parmer VapLock™ Fitting, Yellow PP w/Yellow ETFE, Straight, Compression to Threaded Adapter, 1/8" OD tubing x 1/4-28 UNF(M); 1/Ea
Cole-Parmer | Item: 1202691
Designed for secure liquid-tight connections
₹1,600.00INR / Each
₹1,600.00INR / Each
Buy It Together
Current Item
Cole-Parmer VapLock™ Fitting, Yellow PP w/Yellow ETFE, Straight, Compression to Threaded Adapter, 1/8" OD tubing x 1/4-28 UNF(M); 1/Ea
Cole-Parmer VapLock™ Fitting, Yellow PP w/Yellow ETFE, Straight, Compression to Threaded Adapter, 1/8" OD tubing x 1/4-28 UNF(M); 1/Ea
1202691
₹1,600.00INR / Each
+
Cole-Parmer PTFE Tubing, 1/16" ID x 1/8" OD; 50 Ft
5011945
₹13,300.00INR / Pkg of 1
+ + + + + + + + =
Total Price 53600.00 ADD 10 ITEMS TO CART
Product Description
Key Features
  • Two piece nut and ferrule system
  • Adapter for semi-rigid tubing
  • Color coded for your convenience
More About this Item
Adapters for semi-rigid tubing are color coded by tubing size for easy identification. A two-piece nut and ferrule system provides a liquid-tight connection and withstands pressures up to 500 psi.
Specifications
  • Fitting MaterialPP / ETFE
  • Connection TypeCompression x UNF(M)
  • Thread size UNF(M) (port 2) (InchShort)1/4-28
  • Compression Size / Tube Od (Port 1) (in)1/8
  • Compression Size / Tube Od (Port 1) (mm)3.175
  • ColorYellow / Yellow
  • DescriptionFitting, Yellow Polypropylene, 1/8" OD tubing x 1/4-28 UNF(M), Yellow ETFE Ferrule; 1/EA