Dibafit™ 1/4"-28 UNF(M) Gripper Fittings for 1/16"OD Tubing
One-piece grippers withstand pressures up to 1000 psi!
  • Permanently attached PTFE ferrules are ideal for repeated connect/disconnect
  • Inner PTFE sealing face is the only material in the fluid path
Dibafit gripper fitting system uses a one-piece ferrule that permanently attaches to the outside of semi-rigid tubing. The fitting can spin freely, preventing tubing from twisting. Select from polypropylene (PP), ETFE, or PEEK fittings. PP fittings are available in 10 different colors for easy identification. Mixed pack includes all 10 colors. ETFE and PEEK fittings provide excellent chemical resistance. PEEK fittings withstand higher temperatures and are available in compact or standard head.
1,900.00 - ₹79,000.00INR / Pkg of 10
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
Compareitem specifications
Color Fitting Material Description
Availability Pricing
Dibafit™ Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/16" OD tubing, black; 10/pk WW-21939-60 Mfr # 002100
COMPARE
Black Polypropylene Gripper Fitting, 1/4\"-28 UNF(M) flat bottom, 1/16\" OD tubing, black; 10/pk
₹2,800.00INR / Pkg of 10
Dibafit™ Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/16" OD tubing, brown; 10/pk WW-21939-61 Mfr # 002101
COMPARE
Brown Polypropylene Gripper Fitting, 1/4\"-28 UNF(M) flat bottom, 1/16\" OD tubing, brown; 10/pk
₹1,900.00INR / Pkg of 10
Dibafit™ Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/16" OD tubing, red; 10/pk WW-21939-62 Mfr # 002102
COMPARE
Red Polypropylene Gripper Fitting, 1/4\"-28 UNF(M) flat bottom, 1/16\" OD tubing, red; 10/pk
₹2,800.00INR / Pkg of 10
Dibafit™ Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/16" OD tubing, orange; 10/pk WW-21939-63 Mfr # 002103
COMPARE
Orange Polypropylene Gripper Fitting, 1/4\"-28 UNF(M) flat bottom, 1/16\" OD tubing, orange; 10/pk
₹2,800.00INR / Pkg of 10
Dibafit™ Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/16" OD tubing, yellow; 10/pk WW-21939-64 Mfr # 002104
COMPARE
Yellow Polypropylene Gripper Fitting, 1/4\"-28 UNF(M) flat bottom, 1/16\" OD tubing, yellow; 10/pk
₹2,800.00INR / Pkg of 10
Dibafit™ Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/16" OD tubing, green; 10/pk WW-21939-65 Mfr # 002105
COMPARE
Green Polypropylene Gripper Fitting, 1/4\"-28 UNF(M) flat bottom, 1/16\" OD tubing, green; 10/pk
₹2,800.00INR / Pkg of 10
Dibafit™ Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/16" OD tubing, blue; 10/pk WW-21939-66 Mfr # 002106
COMPARE
Blue Polypropylene Gripper Fitting, 1/4\"-28 UNF(M) flat bottom, 1/16\" OD tubing, blue; 10/pk
₹2,800.00INR / Pkg of 10
Dibafit™ Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/16" OD tubing, violet; 10/pk WW-21939-67 Mfr # 002107
COMPARE
Violet Polypropylene Gripper Fitting, 1/4\"-28 UNF(M) flat bottom, 1/16\" OD tubing, violet; 10/pk
₹2,800.00INR / Pkg of 10
Dibafit™ Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/16" OD tubing, gray; 10/pk WW-21939-68 Mfr # 002108
COMPARE
Gray Polypropylene Gripper Fitting, 1/4\"-28 UNF(M) flat bottom, 1/16\" OD tubing, gray; 10/pk
₹2,800.00INR / Pkg of 10
Dibafit™ Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/16" OD tubing, white; 10/pk WW-21939-69 Mfr # 002109
COMPARE
White Polypropylene Gripper Fitting, 1/4\"-28 UNF(M) flat bottom, 1/16\" OD tubing, white; 10/pk
₹2,800.00INR / Pkg of 10
Dibafit™ Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/16" OD tubing, mixed; 10/pk WW-21939-70 Mfr # 002110
COMPARE
Mixed Polypropylene Gripper Fitting, 1/4\"-28 UNF(M) flat bottom, 1/16\" OD tubing, mixed; 10/pk
₹2,800.00INR / Pkg of 10
Dibafit™ Gripper Fitting, 1/4"-28 UNF(M) flat bottom, 1/16" OD tubing, natural; 10/pk WW-21939-72 Mfr # 002122
COMPARE
Natural PEEK Gripper Fitting, 1/4\"-28 UNF(M) flat bottom, 1/16\" OD tubing, natural; 10/pk
₹7,900.00INR / Pkg of 10
Dibafit™ Gripper Fitting, 1/4"-28 UNF(M), 1/16" OD tubing, natural ETFE; 10/pk WW-21939-74 Mfr # 002130
COMPARE
Natural ETFE Gripper Fitting, 1/4\"-28 UNF(M), 1/16\" OD tubing, natural ETFE; 10/pk
₹79,000.00INR / Pkg of 10
Showing 1- 13 of 13