Cole-Parmer® PTFE Tubing
Chemically inert and resistant to nearly all chemicals and solvents
  • Wide temperature range of –201 to 260°C (–330 to 500°F)
  • Lowest coefficient of friction of any tubing
  • Excellent flexibility
  • Does not contain BPA, phthalates, or PVC
  • Sterilize by autoclave, EtO, and chemical sterilants
  • Complies with FDA 21 CFR 177.1550, USP Class VI, ASTM D 3295
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
A wide operating temperature range, superior chemical resistance, and excellent corrosion resistance make PTFE tubing ideal for use in extreme environments. Smooth, nonstick surface and low permeability characteristics ensure there is no buildup or absorption/adsorption of fluids, minimizing the risk of fluid contamination. PTFE tubing is more flexible than all other tubing in its class, reducing the chances of cracks from bending.
11,000.00 - ₹65,400.00INR / Pkg of 1
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
COMPARE
1/16 1/8 1/32
₹11,000.00INR / Pkg of 1
COMPARE
1/8 3/16 1/32
₹20,400.00INR / Pkg of 1
COMPARE
1/8 1/4 1/16
₹41,300.00INR / Pkg of 1
COMPARE
3/16 1/4 1/32
₹20,700.00INR / Pkg of 1
COMPARE
5/32 1/4 3/64
₹35,800.00INR / Pkg of 1
COMPARE
7/16 1/2 1/32
₹65,400.00INR / Pkg of 1