Cole-Parmer VapLock™ Fitting, Green PP Nut, Blue ETFE Ferrule, Straight, Compression to Threaded Adapter, 1/16" OD x 1/4" NPT(M); 1/EA
Representative image only.
Cole-Parmer VapLock™ Fitting, Green PP Nut, Blue ETFE Ferrule, Straight, Compression to Threaded Adapter, 1/16" OD x 1/4" NPT(M); 1/EA
Cole-Parmer | Item: 1202011
Designed for secure leak-resistant connections
₹2,400.00INR / Each
₹2,400.00INR / Each
Buy It Together
Current Item
Cole-Parmer VapLock™ Fitting, Green PP Nut, Blue ETFE Ferrule, Straight, Compression to Threaded Adapter, 1/16" OD x 1/4" NPT(M); 1/EA
Cole-Parmer VapLock™ Fitting, Green PP Nut, Blue ETFE Ferrule, Straight, Compression to Threaded Adapter, 1/16" OD x 1/4" NPT(M); 1/EA
1202011
₹2,400.00INR / Each
+ + + + + + + + + =
Total Price 82390.00 ADD 10 ITEMS TO CART
Product Description
Key Features
  • Easily identify from a distance with color coded adapters
  • 1/16" OD x 1/4" NPT(M)
  • Green PP nut and blue ETFE ferrule
More About this Item
Adapters for semi-rigid tubing are color coded by tubing size for easy identification. Each connection is comprised of a two-piece nut and ferrule system with polypropylene NPT(M) thread adapter. Wrap NPT thread 3 to 4 times with PTFE tape and wrench tighten for a leak-resistant connection. Adapters withstand pressures up to 500 psi.
Specifications
  • Fitting MaterialPP / ETFE
  • Connection TypeCompression x NPT(M)
  • Thread Size NPT(M) (Port 2) (InchShort)1/4
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Compression Size / Tube Od (Port 1) (mm)1.6
  • ColorGreen nut/blue ferrule/white adapter
  • Max Operating Pressure (PSI)500
  • DescriptionTubing Adapter, 1/16" OD x 1/4" NPT(M), Green PP Nut, Blue ETFE Ferrule; 1/EA