Idex Threaded Adapter with Fitting, Red PEEK, 1/16" ID, 1/4-28 Flat Bottom (F) to 1/8" NPT(M)
Representative image only.
Idex Threaded Adapter with Fitting, Red PEEK, 1/16" ID, 1/4-28 Flat Bottom (F) to 1/8" NPT(M)
Idex | Mfr: U-510| Item: 0201482
Maintain secure connections to 1/8" and 1/4" NPT ports even under high pressure
₹6,100.00INR / Each
₹6,100.00INR / Each
Product Description
Includes
One fitting 02020-93
Key Features
  • Securely join together fluid pathway components
  • Manufactured from PEEK polymer
  • Includes fittings
  • Frit volume 0.6 µL
  • 1 month warranty
  • 1 month warranty
More About this Item
Made from PEEK, these adapters are durable and have excellent chemical resistance. Wrap NPT threads with thread seal tape to ensure leak-free seal.
1 Month Warranty
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Number Of Ports2
  • Thread size UNF(F) (port 1) (InchShort)1/4-28 Flat Bottom
  • Compression Size / Tube Od (Port 1) (in)1/8
  • ColorRed
  • Max Operating Pressure (PSI)500
  • Orifice Dimensions (in)1/16
  • DescriptionThreaded Adapter with Fitting, Red PEEK, 1/16" ID, 1/4-28 Flat Bottom (F) to 1/8" NPT(M)