Cole-Parmer VapLock™ Fitting, Polypropylene, Straight, Compression to Threaded Adapter, 3/8" OD Tubing x 1/2" NPT(M); 1/EA
Representative image only.
Cole-Parmer VapLock™ Fitting, Polypropylene, Straight, Compression to Threaded Adapter, 3/8" OD Tubing x 1/2" NPT(M); 1/EA
Cole-Parmer | Item: 1202657
Designed for secure leak-resistant connections
₹570.00INR / Each
₹570.00INR / Each
Buy It Together
Current Item
Cole-Parmer VapLock™ Fitting, Polypropylene, Straight, Compression to Threaded Adapter, 3/8" OD Tubing x 1/2" NPT(M); 1/EA
Cole-Parmer VapLock™ Fitting, Polypropylene, Straight, Compression to Threaded Adapter, 3/8" OD Tubing x 1/2" NPT(M); 1/EA
1202657
₹570.00INR / Each
+ + + + =
Total Price 29470.00 ADD 5 ITEMS TO CART
Product Description
Key Features
  • Constructed of durable polypropylene
More About this Item
Adapters with large bore for semi-rigid tubing are constructed of two-piece molded polypropylene components and do not include a separate ferrule. Adapters withstand pressures up to 100 psi. Wrap NPT threads 3 to 4 times with PTFE tape and wrench tighten for leak-resistant connections.
Specifications
  • Fitting MaterialPolypropylene
  • Connection TypeCompression x NPT(M)
  • Thread Size NPT(M) (Port 2) (InchShort)1/2
  • Compression Size / Tube Od (Port 1) (in)3/8
  • Compression Size / Tube Od (Port 1) (mm)9.5
  • DescriptionFitting, Polypropylene, Straight, Compression to Threaded Adapter, 3/8" OD Tubing x 1/2" NPT(M); 1/EA