Cole-Parmer VapLock™ Fitting, Black PP w/ White PTFE, Straight, Compression to Threaded Adapter, 2.2 mm OD x 1/4-28 UNF(M); 10/PK
Representative image only.
Cole-Parmer VapLock™ Fitting, Black PP w/ White PTFE, Straight, Compression to Threaded Adapter, 2.2 mm OD x 1/4-28 UNF(M); 10/PK
Cole-Parmer | Item: 1202005
Designed for secure liquid-tight connections
₹8,500.00INR / Pkg of 10
₹8,500.00INR / Pkg of 10
Buy It Together
Current Item
Cole-Parmer VapLock™ Fitting, Black PP w/ White PTFE, Straight, Compression to Threaded Adapter, 2.2 mm OD x 1/4-28 UNF(M); 10/PK
Cole-Parmer VapLock™ Fitting, Black PP w/ White PTFE, Straight, Compression to Threaded Adapter, 2.2 mm OD x 1/4-28 UNF(M); 10/PK
1202005
₹8,500.00INR / Pkg of 10
+
Cole-Parmer PTFE Tubing, 1/16" ID x 1/8" OD; 50 Ft
5011945
₹13,300.00INR / Pkg of 1
+ + + + + + + + =
Total Price 57100.00 ADD 10 ITEMS TO CART
Product Description
Key Features
  • Two piece nut and ferrule system
  • Adapter for semi-rigid tubing
  • Color coded for your convenience
More About this Item
Adapters for semi-rigid tubing are color coded by tubing size for easy identification. A two-piece nut and ferrule system provides a liquid-tight connection and withstands pressures up to 500 psi.
Specifications
  • Fitting MaterialPP / PTFE
  • Connection TypeCompression x UNF(M)
  • Thread size UNF(M) (port 2) (InchShort)1/4-28
  • Compression Size / Tube Od (Port 1) (in)3/32
  • Compression Size / Tube Od (Port 1) (mm)2.2
  • ColorBlack / White
  • DescriptionTubing Adapter, 2.2 mm OD x 1/4-28 UNF(M), black PP w/ white ETFE; 10/PK