Cole-Parmer Support Stand, (9"L x 5-1/2" base) with 24"H x 1/2" dia. stainless steel rod
Representative image only.
Cole-Parmer Support Stand, (9"L x 5-1/2" base) with 24"H x 1/2" dia. stainless steel rod
Cole-Parmer | Item: 0471292
Enhance the performance of your ultrasonic processor
₹18,300.00INR / Each
₹18,300.00INR / Each
Buy It Together
Current Item
Cole-Parmer Support Stand, (9"L x 5-1/2" base) with 24"H x 1/2" dia. stainless steel rod
Cole-Parmer Support Stand, (9"L x 5-1/2" base) with 24"H x 1/2" dia. stainless steel rod
0471292
₹18,300.00INR / Each
+ + + +
Cole-Parmer Coupler for 500- and 750-Watt Processors
0471085
₹56,600.00INR / Each
+ + + + + =
Total Price 465900.00 ADD 10 ITEMS TO CART
Product Description
Key Features
  • Compatible with Cole-Parmer 500- and 700-Watt ultrasonic processors
  • Durable cast-iron base and stainless steel rod for harsh applications
  • 3 year warranty
More About this Item
Choose from a wide range of accessories including support stands and converter clamps for your Cole-Parmer 500- and 700-Watt ultrasonic processors.
Optional stand has black enameled cast-iron base and zinc-plated rod. Base measures 9"L x 5-1/2"W (229 x 140 mm). Rod measures 24"H (609 mm) x 1/2"dia (12.6 mm).
Order converter clamp 04714-90 for use with 130-watt ultrasonic processors.
Order light-duty converter clamp 04714-91 for use with 500- and 750-watt ultrasonic processors.
For industrial applications with 500- and 750-watt ultrasonic processors, order heavy-duty converter clamp 04710-94.
Order all converter clamps separately at right.
3 Year Warranty
Specifications
  • Compatible WithCole-Parmer 500 -700 watt ultrasonic processors
  • DescriptionSupport Stand, (9"L x 5-1/2" base) with 24"H x 1/2" dia. stainless steel rod