Cole-Parmer Coupler for 500- and 750-Watt Processors
Representative image only.
Cole-Parmer Coupler for 500- and 750-Watt Processors
Cole-Parmer | Item: 0471085
₹56,600.00INR / Each
₹56,600.00INR / Each
Buy It Together
Current Item
Cole-Parmer Coupler for 500- and 750-Watt Processors
Cole-Parmer Coupler for 500- and 750-Watt Processors
0471085
₹56,600.00INR / Each
+
Cole-Parmer Ultrasonic Probe/Horn, Solid; 1/2" Dia
0471051
₹111,000.00INR / Each
+ + + + + =
Total Price 448500.00 ADD 7 ITEMS TO CART
Product Description
Key Features
  • 1 year warranty
More About this Item
Probes are made of titanium and are autoclavable. Optional 2-mm probe fits into microcentrifuge tubes; use to process small-volume samples.
Use this coupler to attach stepped tip 04712-12 to 500- and 750-watt ultrasonic processors.
Coupler for 500- and 750-Watt Ultrasonic Processors
1 Year Warranty
Specifications
  • DescriptionCoupler for 500- and 750-Watt Processors