Cole-Parmer Ultrasonic Processor Stainless Steel Temperature Probe
Representative image only.
Cole-Parmer Ultrasonic Processor Stainless Steel Temperature Probe
Cole-Parmer | Item: 0471150
₹23,000.00INR / Each
₹23,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer Ultrasonic Processor Stainless Steel Temperature Probe
Cole-Parmer Ultrasonic Processor Stainless Steel Temperature Probe
0471150
₹23,000.00INR / Each
+ + + +
Cole-Parmer Coupler for 500- and 750-Watt Processors
0471085
₹56,600.00INR / Each
+ + =
Total Price 454500.00 ADD 7 ITEMS TO CART
Product Description
Key Features
  • 3 year warranty
More About this Item
The temperature probe used in conjunction with ultrasonic processors with temperature controller prevents harmful overheating of the sample by terminating the ultrasonic when the sample reaches a predetermined temperature limit.
Use probe with 500- and 750-watt ultrasonic processors 04711-30, 04711-35, 04711-40, and 04711-45.
3 Year Warranty
Specifications
  • DescriptionUltrasonic Processor Stainless Steel Temperature Probe