Cole-Parmer® Aluminum Apparatus Positioner/Lab Jack
Safely position samples or equipment
  • Excellent side-to-side rigidity and support
These laboratory scissor jacks provide stable, height-adjustable support for hot plates, baths, glassware, and many other items. Positioners are constructed of rugged, chemical-resistant anodized aluminum with stainless steel operating screw. Top-plate skid pad prevents slipping.
Each positioner is precision engineered for smooth, continuous operation throughout its range. Easily set the exact height needed with the turn of a knob.
28,000.00 - ₹120,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (lbs) Size Description
Availability Pricing
COMPARE
66 Lift height: 2.5 to 5" Lab Jack, 4" x 4" Top, 66 Lb Capacity
₹28,000.00INR / Each
COMPARE
132 Lift height: 3 to 9.75" Lab Jack, 6" x 6" Top, 132 lb Capacity
₹37,600.00INR / Each
COMPARE
176 Lift height: 3 to 9.75" Lab Jack, 8" x 8" Top, 176 Lb Capacity
₹41,200.00INR / Each
COMPARE
186 Lift height: 3.5 to 13" Lab Jack, 10" x 10" Top, 186 Lb Capacity
₹120,000.00INR / Each