Cole-Parmer Glass Media Bottles, Class A, Round, 100 mL (3.4 oz); 10/Pk

Representative image only.

Cole-Parmer Glass Media Bottles, Class A, Round, 100 mL (3.4 oz); 10/Pk

Cole-Parmer | Item: 3450338

Class A round glass media storage bottles ideal for mixing, sampling, and storing mixtures, reagents, solvents, and solutions

₹12,100.00INR / Case of 10

₹12,100.00INR / Case of 10

Buy It Together

Current Item

Cole-Parmer Glass Media Bottles, Class A, Round, 100 mL (3.4 oz); 10/Pk
Cole-Parmer Glass Media Bottles, Class A, Round, 100 mL (3.4 oz); 10/Pk
3450338

₹12,100.00INR / Case of 10
+
Cole-Parmer Narrow-Mouth Plastic Bottle, PPCO, 60mL; 12/PK
0603426

₹2,700.00INR / Pkg of 12
+ +
Cole-Parmer Wide-Mouth Plastic Bottle, HDPE, 4000mL
0605520

₹6,200.00INR / Each
+
Cole-Parmer Wide-Mouth Plastic Bottle, PP, 2000mL (64 oz)
0605710

₹5,000.00INR / Each
+
Cole-Parmer 7981 Graduated Round Bottle, Clear Glass, 30mL; 48/CS
3460110

₹39,100.00INR / Case of 48
+ +
Cole-Parmer Graduated Round Bottle, Clear Glass, 250mL; 24/CS
3480046

₹25,000.00INR / Case of 24
+ + =
Total Price 130300.00 ADD 10 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Polypropylene cap.

Key Features

  • Wide-mouth opening for easy pouring
  • Resistant to corrosion by most chemicals
  • Withstand temperatures from -50 to 550 °C (-58 to 1022 °F)
  • Autoclavable PP cap and bottle
  • 5 year warranty

More About this Item

Made of borosilicate 3.3 glass, these Class A, round media bottles provide outstanding corrosion resistance and meet ASTM E438 requirements. Able to withstand temperatures from -50 to 550 °C (-58 to 1022 °F), they have a wide mouth opening that allows for easy pouring. The clear, colorless walls have a marking spot for labeling and graduations showing approximate volume. Both the bottle and polypropylene cap are autoclavable.

5 Year Warranty
Specifications
  • Capacity (mL)100
  • MaterialBorosilicate Glass
  • Min Graduation (mL)20
  • Max Graduation (mL)80
  • Graduation Interval (mL)20
  • Cap MaterialPolypropylene (PP)
  • Cap / Stopper Size45 mm, GL45
  • Cap ColorBlue
  • Bottle ColorClear
  • AutoclavableYes
  • Withstands High Temperature (DegreeF)1022
  • Withstands Low Temperature (° F)-58
  • Neck Inside Diameter (ID) (mm)32
  • Neck Inside Diameter (ID) (in)1 1/4
  • Outside Diameter (OD) (in)2 13/64
  • Outside Diameter (OD) (cm)5.6
  • Overall Height (in)3 31/32
  • Overall Height (cm)10.1
  • DescriptionGlass Media Bottles, Class A, Round, 100mL (3.4 oz); 10/PK
  • Withstands High Temperature (DegreeC)550
  • Withstands Low Temperature (DegreeC)-50
  • Warranty5 years
Reviews
Q&A