Cole-Parmer® Economy Wide-Mouth Plastic Bottles, HDPE
Light-weight and flexible for general-purpose laboratory applications
  • Withstands freezer storage down to -100°C
  • Translucent and more rigid than LDPE
  • Uniform walls resist splitting and punctures
  • Meets USP Class VI requirements
These wide-mouth HDPE bottles are easy to fill with dry or liquid materials and resistant to most chemicals. May be used for freezer storage down to -148°F (-100°C). Suitable for packing, storage, and shipping liquids. All have leakproof polypropylene screw caps.
2,000.00 - ₹42,000.00INR / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
COMPARE
30 28 mm High Density Polyethyene (HDPE)
₹2,000.00INR / Pkg of 12
COMPARE
30 28 mm High Density Polyethyene (HDPE)
₹21,000.00INR / Pkg of 72
COMPARE
60 28 mm High Density Polyethyene (HDPE)
₹2,300.00INR / Pkg of 12
COMPARE
60 28 mm High Density Polyethyene (HDPE)
₹23,000.00INR / Pkg of 72
COMPARE
2000 100 mm High Density Polyethyene (HDPE)
₹2,300.00INR / Each
COMPARE
125 38 mm High Density Polyethyene (HDPE)
₹6,000.00INR / Pkg of 12
COMPARE
125 38 mm High Density Polyethyene (HDPE)
₹30,000.00INR / Pkg of 72
COMPARE
250 43 mm High Density Polyethyene (HDPE)
₹4,000.00INR / Pkg of 12
COMPARE
250 43 mm High Density Polyethyene (HDPE)
₹22,500.00INR / Pkg of 72
COMPARE
500 53 mm High Density Polyethyene (HDPE)
₹5,800.00INR / Pkg of 12
COMPARE
500 53 mm High Density Polyethyene (HDPE)
₹42,000.00INR / Pkg of 48
COMPARE
1000 63 mm High Density Polyethyene (HDPE)
₹6,300.00INR / Pkg of 6
COMPARE
1000 63 mm High Density Polyethyene (HDPE)
₹23,400.00INR / Pkg of 24
Showing 1- 13 of 13