Cole-Parmer FD 600 Benchtop Freeze Dryer
Representative image only.
Cole-Parmer FD 600 Benchtop Freeze Dryer
Cole-Parmer | Item: 1571701
An all-in-one, intelligent benchtop laboratory freeze dryer that combines automated, hands-off operation with full manual control for consistent, high-quality results
₹3,599,000.00INR / Each
₹3,599,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer FD 600 Benchtop Freeze Dryer
Cole-Parmer FD 600 Benchtop Freeze Dryer
1571701
₹3,599,000.00INR / Each
+
Pfeiffer HiScroll 6 Scroll Vacuum Pump
1571702
₹2,297,000.00INR / Each
=
Total Price 5896000.00 ADD 2 ITEMS TO CART
Product Description
Includes
Five shelf stack with 5 stainless steel trays.
Key Features
  • AutoDry technology automatically detects the end of the drying cycle, optimizing efficiency and eliminating manual intervention
  • Integrated freezing and drying prevent sample transfer issues, improving product quality and process reliability
  • Designed for routine use in Environmental, Molecular and Cell Biology, Diagnostics, Wound Care and General Lab applications
  • Temperature-controlled, electrically heated shelves enable precise process control and can reduce drying time by up to 50%
  • Front-loading, easy-access design with interchangeable shelf configurations simplifies setup and supports diverse applications
More About this Item
The Cole-Parmer® FD 600 benchtop freeze dryer is a versatile tray freeze dryer designed for both new and experienced users who demand reliability, flexibility, and ease of use. Its all-in-one process freezes and dries samples within a single system, eliminating the risks of ice crystal damage and product thawing associated with external freezing and sample transfer. With AutoDry mode, the FD 600 continuously monitors process conditions using an adaptive algorithm, adjusting parameters in real time and automatically stopping the cycle at the optimal endpoint for consistent, reproducible results.
For users who prefer full control, the FD 600 also offers manual operation, vacuum oven mode, and an intuitive touchscreen interface with 12 programmable, nameable recipes—six preloaded for common applications. Five electrically heated shelves and stainless steel trays are included as standard, supporting an ice capacity of up to 8 kg and removal of up to 4 kg per 24 hours. Designed for modern laboratories, the FD 600 features power recovery protection, energy-efficient components, and an ozone-friendly natural refrigerant, delivering lower operating costs and a reduced environmental footprint. (Vacuum pump sold separately.)
Specifications
  • Capacity (Liters)8
  • Width (InchShort)29 29/64
  • Cooling Capacity (° F)-49
  • Height (InchShort)19 61/64
  • Cooling Capacity (° C)-45
  • Sublimation Rate (Liters/day)4
  • Power (VAC)120 / 240
  • Power (Hz)50/60
  • DescriptionBenchtop Freeze Dryer
  • Warranty2 years