Cole-Parmer FD 600 Three Shelf Stack
Representative image only.
Cole-Parmer FD 600 Three Shelf Stack
Cole-Parmer | Item: 1571706
Heated shelf stacks and trays designed to expand the capacity, flexibility, and performance of the FD 600 freeze dryer
₹524,000.00INR / Each
₹524,000.00INR / Each
Product Description
Key Features
  • Fixed heated shelf stacks from 1 to 9 shelves provide configurable capacity for thin liquids, bulk samples, or tall labware
  • Temperature-controlled shelves ensure uniform heat transfer and consistent drying across all shelf configurations
  • Stainless steel trays in multiple depths support a wide range of product formats and application needs
  • 3 heated shelves for use with labware or trays up to 57 mm tall
More About this Item
The FD 600 accessory lineup is engineered to maximize the versatility and efficiency of your freeze-drying workflow.
To support diverse sample types and throughput requirements, the FD 600 fixed heated shelf stacks are available in configurations ranging from 1 to 9 shelves. Smaller inter-shelf spacing enables high-capacity drying of thin liquids, while larger spacing accommodates bulk products, deep well plates, and tall containers. All shelves are temperature controlled for uniform drying and long-term durability, and interchangeable stainless steel trays in 12 mm, 15 mm, and 20 mm depths allow users to tailor the system to their exact application. Together, these accessories transform the FD 600 into a highly adaptable freeze-drying platform suitable for research, development, and production-scale laboratory work.
The FD 600 Fixed Three Heated Shelf Stack is specifically designed to accommodate larger-format products, including deep well plates and bulk containers. It features three fixed, temperature-controlled shelves, each measuring 465 mm x 210 mm, delivering a total usable shelf area of 2,928 cm².
With an inter-shelf distance of 59 mm, this configuration provides ample vertical space and ensures optimal airflow for effective freeze drying. Durable and easy to maintain, the 3-shelf stack is ideal for applications requiring larger product volumes or specialized formats.
Specifications
  • Compatible WithCole-Parmer® FD 600 Benchtop Freeze Dryer
  • DescriptionThree Shelf Stack