Cole-Parmer Stainless Steel Trays for Five Shelf Stack, 5/Pk
Representative image only.
Cole-Parmer Stainless Steel Trays for Five Shelf Stack, 5/Pk
Cole-Parmer | Item: 1571710
Heated shelf stacks and trays designed to expand the capacity, flexibility, and performance of the FD 600 freeze dryer
₹88,600.00INR / Pkg of 5
₹88,600.00INR / Pkg of 5
Product Description
Key Features
  • Fixed heated shelf stacks from 1 to 9 shelves provide configurable capacity for thin liquids, bulk samples, or tall labware
  • Temperature-controlled shelves ensure uniform heat transfer and consistent drying across all shelf configurations
  • Stainless steel trays in multiple depths support a wide range of product formats and application needs
  • 200 W x 450 L x 20 H mm
  • For use with the FD 600 Five Shelf Stack
More About this Item
The FD 600 accessory lineup is engineered to maximize the versatility and efficiency of your freeze-drying workflow.
To support diverse sample types and throughput requirements, the FD 600 fixed heated shelf stacks are available in configurations ranging from 1 to 9 shelves. Smaller inter-shelf spacing enables high-capacity drying of thin liquids, while larger spacing accommodates bulk products, deep well plates, and tall containers. All shelves are temperature controlled for uniform drying and long-term durability, and interchangeable stainless steel trays in 12 mm, 15 mm, and 20 mm depths allow users to tailor the system to their exact application. Together, these accessories transform the FD 600 into a highly adaptable freeze-drying platform suitable for research, development, and production-scale laboratory work.
Specifications
  • MaterialStainless steel
  • Compatible WithCole-Parmer® FD 600 Benchtop Freeze Dryer
  • DescriptionStainless Steel Trays for Five Shelf Stack, 5/Pk