Cole-Parmer FD 600 Seven Shelf Stack
Representative image only.
Cole-Parmer FD 600 Seven Shelf Stack
Cole-Parmer | Item: 1571704
Heated shelf stacks and trays designed to expand the capacity, flexibility, and performance of the FD 600 freeze dryer
₹663,000.00INR / Each
₹663,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer FD 600 Seven Shelf Stack
Cole-Parmer FD 600 Seven Shelf Stack
1571704
₹663,000.00INR / Each
+
Cole-Parmer Stainless Steel Trays for Seven Shelf Stack, 7/Pk
1571709
₹117,000.00INR / Pkg of 7
=
Total Price 780000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Fixed heated shelf stacks from 1 to 9 shelves provide configurable capacity for thin liquids, bulk samples, or tall labware
  • Temperature-controlled shelves ensure uniform heat transfer and consistent drying across all shelf configurations
  • Stainless steel trays in multiple depths support a wide range of product formats and application needs
  • 7 heated shelves for use with labware or trays up to 20 mm tall
More About this Item
The FD 600 accessory lineup is engineered to maximize the versatility and efficiency of your freeze-drying workflow.
To support diverse sample types and throughput requirements, the FD 600 fixed heated shelf stacks are available in configurations ranging from 1 to 9 shelves. Smaller inter-shelf spacing enables high-capacity drying of thin liquids, while larger spacing accommodates bulk products, deep well plates, and tall containers. All shelves are temperature controlled for uniform drying and long-term durability, and interchangeable stainless steel trays in 12 mm, 15 mm, and 20 mm depths allow users to tailor the system to their exact application. Together, these accessories transform the FD 600 into a highly adaptable freeze-drying platform suitable for research, development, and production-scale laboratory work.
Designed for high-capacity processing of smaller products, the FD 600 Fixed Seven Heated Shelf Stack features seven fixed, temperature-controlled shelves measuring 465 mm x 210 mm, offering a total usable shelf area of 6,832 cm².
With an inter-shelf distance of 22 mm, this configuration supports excellent airflow and consistent drying performance throughout the chamber. Built for durability and easy cleaning, it is ideal for demanding freeze-drying applications.
Requires use of 12 mm or 15 mm trays (not included).
Specifications
  • Compatible WithCole-Parmer® FD 600 Benchtop Freeze Dryer
  • DescriptionSeven Shelf Stack