Cole-Parmer EX Adjustable Volume Pipette, 0.5-10.0 uL; Each
Representative image only.
Cole-Parmer EX Adjustable Volume Pipette, 0.5-10.0 uL; Each
Cole-Parmer | Item: 0790903
Autoclavability and enhanced UV resistance provides added protection from biohazardous elements
₹64,300.00INR / Each
for an additional
₹14,500.00INR / Each
for an additional
₹25,800.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹64,300.00INR / Each
Buy It Together
Current Item
Cole-Parmer EX Adjustable Volume Pipette, 0.5-10.0 uL; Each
Cole-Parmer EX Adjustable Volume Pipette, 0.5-10.0 uL; Each
0790903
₹64,300.00INR / Each
+
Cole-Parmer Macropipettor Pipette Tips, 0.4-2 mL, Clear; 200/Bag
0785922
₹11,300.00INR / Bag of 200
+
Cole-Parmer Pipette Tips, 1-10 mL, White; 200/Pk
0790925
₹14,000.00INR / Bag of 200
+ + + +
Cole-Parmer Pipette Tip, 0.1 to 10 µL, Bulk; 1000/BG
2450142
₹4,500.00INR / Bag of 1000
+ +
Cole-Parmer Long Pipette Tip, 20 to 200 µL, Bulk; 1000/BG
2450243
₹4,900.00INR / Bag of 1000
+
Cole-Parmer Pipette Tip, 100 to 1000 µL, Bulk; 1000/BG
2450261
₹5,700.00INR / Bag of 1000
=
Total Price 198400.00 ADD 10 ITEMS TO CART
Customers Also Bought
Product Description
Includes
calibration document supplied by the manufacturer. Order tips separately.
Key Features
  • Locking lever prevents accidental volume change
  • Self-lubricating PTFE seal-ring eliminates routine lubrication
  • Sterilize by autoclave or 254 nm UV light
  • 2 year warranty
More About this Item
This lightweight, slender pipettor features molded channels to improve grip and its smooth plunger action reduces operator fatigue. Easily adjust the pipette volume by turning the colored dial, and lock dial feature prevents accidental volume changes during use. Separate tip release button eliminates accidental release.
Two year warranty
Specifications
  • SterileNo
  • AutoclavableYes
  • Max Volume (µL)10
  • Min Volume (µL)0.5
  • Min Volume (mL)0.0005
  • Max Volume (mL)0.01
  • Min Dispensing (mL)0.0005
  • Max Dispensing (mL)0.01
  • Increments (µL)0.01
  • Accuracy (%)±5.00% to ±1.00%
  • Number Of Channels1
  • Tip EjectorYes
  • Body MaterialStainless Steel (SS) and polypropylene (PP) plunger assembly
  • DescriptionAdjustable Volume Pipette, 0.5-10.0 uL; Each