Cole-Parmer® EX Adjustable-Volume Pipettors
Autoclavability and enhanced UV resistance provides added protection from biohazardous elements
  • Locking lever prevents accidental volume change
  • Self-lubricating PTFE seal-ring eliminates routine lubrication
  • Sterilize by autoclave or 254 nm UV light
This lightweight, slender pipettor features molded channels to improve grip and its smooth plunger action reduces operator fatigue. Easily adjust the pipette volume by turning the colored dial, and lock dial feature prevents accidental volume changes during use. Separate tip release button eliminates accidental release.
46,500.00 - ₹50,900.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Autoclavable Max Volume (mL) Min Volume (mL)
Availability Pricing
COMPARE
Yes 0.002 0.0001
₹48,000.00INR / Each
COMPARE
Yes 0.01 0.0005
₹46,500.00INR / Each
COMPARE
Yes 0.02 0.002
₹46,900.00INR / Each
COMPARE
Yes 0.1 0.01
₹50,900.00INR / Each
COMPARE
Yes 0.2 0.02
₹46,900.00INR / Each
COMPARE
Yes 1 0.1
₹46,800.00INR / Each
COMPARE
Yes 5 1
₹49,000.00INR / Each
COMPARE
Yes 10 1
₹46,800.00INR / Each
Showing 1- 8 of 8