Cole-Parmer Pipette Tip, 0.1 to 10 µL, 10 Racks; 960/PK
Representative image only.
Cole-Parmer Pipette Tip, 0.1 to 10 µL, 10 Racks; 960/PK
Cole-Parmer | Item: 2450148
Tips free of human touch is purity at its best
₹2,900.00INR / Pkg of 960
₹2,900.00INR / Pkg of 960
Buy It Together
Current Item
Cole-Parmer Pipette Tip, 0.1 to 10 µL, 10 Racks; 960/PK
Cole-Parmer Pipette Tip, 0.1 to 10 µL, 10 Racks; 960/PK
2450148
₹2,900.00INR / Pkg of 960
+
Cole-Parmer Adjustable-Volume Pipettor, 500 to 5000 µL
2450109
₹8,400.00INR / Each
=
Total Price 11300.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Tip Options: Standard and filtered
  • Specially developed PE filters free of chemical additives stops aerosols and contaminations
  • Tips also leach-free, ensuring maximum recovery of sample yield
  • Volume measurements, along the tube are marked in increments for easy identification
  • Produced from Swiss-made Diamond Finish Mold
  • Made of FDA approved special medical grade virgin polypropylene plastic with high transparency
  • Unique label coding in packaging for easy identification
  • Nonsterile, nonfilter tips Volume: 0.1 to 10 µL
More About this Item
Not your everyday graduated pipette tips. It starts with a touch-free manufacturing facility and stringent quality control process to make these clear, plastic tips certified free of any affecting biological elements, such as human DNA, DNase and RNase and Endotoxin, or Pyrogen and PCR inhibitors. They also are noncytotoxic.
It’s about their innovative design, too. The profile and dimensions of the tips are engineered to accurately measure and transfer a volume of liquid from one container to another, as well as adapt to a wide-range of pipettes.
It ends with preserving their purity in the following packaging options, which are free of lubricants and heavy metals.
Rack Pack: Five packs of tips with a variety of options for the number of tips in each pack. This choice comes with an autoclave container with stackable and color-coded trays for easy identification.
Bio DNase-FreeNonpyrogenicRNase-Free
Specifications
  • SterileNo
  • Max Volume (µL)10
  • Min Volume (µL)0.1
  • Tip StyleStandard, 3.13 cm L
  • Body MaterialPolypropylene (PP)
  • Filter TypeNone
  • Package OfStandard Racks, 10 x 96
  • DescriptionPipette Tip, 0.1 to 10 µL, 10 Racks; 960/PK