Cole-Parmer Cell Strainer, Sterile, 100 µm, Yellow, Individually Wrapped; 50/Cs
Representative image only.
Cole-Parmer Cell Strainer, Sterile, 100 µm, Yellow, Individually Wrapped; 50/Cs
Cole-Parmer | Item: 0633671
Quickly strain, isolate, and detach cells
₹36,200.00INR / Case of 50
₹36,200.00INR / Case of 50
Buy It Together
Current Item
Cole-Parmer Cell Strainer, Sterile, 100 µm, Yellow, Individually Wrapped; 50/Cs
Cole-Parmer Cell Strainer, Sterile, 100 µm, Yellow, Individually Wrapped; 50/Cs
0633671
₹36,200.00INR / Case of 50
+ + =
Total Price 87300.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Nylon mesh produces consistent, reproducible results
  • Color-coded frame for easy identification of mesh porosity
  • Designed to be inserted into top of 50-mL centrifuge tube
  • Extended gripping tab makes handling easy
  • Gamma irradiated
  • 100 µm mesh pore size
  • Individually wrapped
More About this Item
Cole-Parmer® Essentials cell strainers are sterile, easy-to-use devices for isolating primary cells to consistently obtain a uniform single-cell suspension from tissues. Constructed of a durable polypropylene housing with nylon mesh filter, these cell strainers provide a faster, easier alternative to gauze or paper filtration. The elongated lip on the housing enable aseptic handling with forceps. Sterile strainers are gamma irradiated to eliminate the risk of cross contamination.
Order convex head pestle with molded textured surface to better manipulate material in the strainer. Pestle sold separately.
Sterile
Specifications
  • Membrane MaterialNylon
  • Pore Size (micron)100
  • Housing MaterialPolypropylene
  • SterileYes
  • DescriptionCell Strainer, Sterile, 100 µm, Yellow, Individually Wrapped; 50/Cs
  • ColorYellow