Cole-Parmer® Cell Strainers, Sterile
Quickly strain, isolate, and detach cells
  • Nylon mesh produces consistent, reproducible results
  • Color-coded frame for easy identification of mesh porosity
  • Designed to be inserted into top of 50-mL centrifuge tube
  • Extended gripping tab makes handling easy
  • Gamma irradiated
Cole-Parmer® Essentials cell strainers are sterile, easy-to-use devices for isolating primary cells to consistently obtain a uniform single-cell suspension from tissues. Constructed of a durable polypropylene housing with nylon mesh filter, these cell strainers provide a faster, easier alternative to gauze or paper filtration. The elongated lip on the housing enable aseptic handling with forceps. Sterile strainers are gamma irradiated to eliminate the risk of cross contamination.
Order convex head pestle with molded textured surface to better manipulate material in the strainer. Pestle sold separately.
31,200.00 - ₹36,200.00INR / Case of 50
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Pore Size (micron) Description
Availability Pricing
COMPARE
Blue 40 Cell Strainer, Sterile, 40 µm, Blue, Individually Wrapped; 50/Cs
₹36,200.00INR / Case of 50
COMPARE
Blue 40 Cell Strainer, Sterile, 40 µm, Blue, Bulk Packed; 50/CS
₹34,700.00INR / Case of 50
COMPARE
White 70 Cell Strainer, Sterile, 70 µm, White, Individually Wrapped; 50/Cs
₹33,300.00INR / Case of 50
COMPARE
White 70 Cell Strainer, Sterile, 70 µm, White, Bulk Packed; 50/Cs
₹34,700.00INR / Case of 50
COMPARE
Yellow 100 Cell Strainer, Sterile, 100 µm, Yellow, Individually Wrapped; 50/Cs
₹36,200.00INR / Case of 50
COMPARE
Yellow 100 Cell Strainer, Sterile, 100 µm, Yellow, Bulk Packed; 50/Cs
₹31,200.00INR / Case of 50